5-Chloro-2-methyl-4-nitroaniline 98%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-4-chloro-5-nitrotoluene

OR42207-25G OR42207-5G
APOSOR42207-25GEA 81.2 EUR
APOSOR42207-25G APOSOR42207-5G
5-Chloro-2-methyl-4-nitroaniline 98%
5-Chloro-2-methyl-4-nitroaniline
Formel: C₇H₇ClN₂O₂
Molecular Weight: 186.596 g/mol
Schmelzpunkt: 164…167 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00043921
CAS-Nummer: 13852-51-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR