4-(Cyanoacetyl)piperidine, N-BOC protected 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-(2-cyanoacetyl)piperidine-1-carboxylate, tert-Butyl 4-(cyanoacetyl)piperidine-1-carboxylate, 3-[1-(tert-Butoxycarbonyl)piperidin-4-yl]-3-oxopropanenitrile, 1-(tert-Butoxycarbonyl)-4-(cyanoacetyl)piperidine

OR42212-1G OR42212-250MG
APOSOR42212-1GEA 43.5 EUR
APOSOR42212-1G APOSOR42212-250MG
4-(Cyanoacetyl)piperidine, N-BOC protected 97%
4-(Cyanoacetyl)piperidine, N-BOC protected
Formel: C₁₃H₂₀N₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD08059465
CAS-Nummer: 660406-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR