4-[5-(But-1-yl)-1,3,4-oxadiazol-2-yl]benzaldehyde

Lieferant: Apollo Scientific
Achtung

OR42228-1G OR42228-250MG
APOSOR42228-1GEA 198 EUR
APOSOR42228-1G APOSOR42228-250MG
4-[5-(But-1-yl)-1,3,4-oxadiazol-2-yl]benzaldehyde
4-[5-(But-1-yl)-1,3,4-oxadiazol-2-yl]benzaldehyde
Formel: C₁₃H₁₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18909698
CAS-Nummer: 1061672-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR