8-Methoxyquinoline-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carboxy-8-methoxyquinoline

OR42240-1G OR42240-250MG
APOSOR42240-1GEA 314 EUR
APOSOR42240-1G APOSOR42240-250MG
8-Methoxyquinoline-3-carboxylic acid
8-Methoxyquinoline-3-carboxylic acid
Formel: C₁₁H₉NO₃
Molecular Weight: 203,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375327
CAS-Nummer: 71082-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR