4-Bromo-1-cyclopropyl-1,3-dihydro-2H-benzimidazol-2-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-1-cyclopropyl-1H-benzo[d]imidazol-2(3H)-one
OR42259-1G
OR42259-500MG
APOSOR42259-1GEA
647
EUR
APOSOR42259-1G
APOSOR42259-500MG
4-Bromo-1-cyclopropyl-1,3-dihydro-2H-benzimidazol-2-one
4-Bromo-1-cyclopropyl-1,3-dihydro-2H-benzimidazol-2-one
Formel:
C₁₀H₉BrN₂O Molecular Weight: 253,095 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22689777 CAS-Nummer: 1416714-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...