3-Hydroxyquinoline-4-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR42261-100MG OR42261-250MG
APOSOR42261-100MGEA 111 EUR
APOSOR42261-100MG APOSOR42261-250MG
3-Hydroxyquinoline-4-carbonitrile
3-Hydroxyquinoline-4-carbonitrile
Formel: C₁₀H₆N₂O
Molecular Weight: 170,167 g/mol
Schmelzpunkt: 201…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18415946
CAS-Nummer: 100517-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR