Methyl 5-iodo-2-methylbenzoate 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR42276-1G
OR42276-25G
OR42276-5G
APOSOR42276-1GEA
21.9
EUR
APOSOR42276-1G
APOSOR42276-25G
APOSOR42276-5G
Methyl 5-iodo-2-methylbenzoate 95%
Methyl 5-iodo-2-methylbenzoate
Formel:
C₉H₉IO₂ Molecular Weight: 276,071 g/mol Schmelzpunkt: 60 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD11112847 CAS-Nummer: 103440-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...