Methyl 6-amino-4-methylnicotinate
Lieferant: Apollo Scientific
|
Achtung
|
OR42282-1G
OR42282-250MG
APOSOR42282-1GEA
522
EUR
APOSOR42282-1G
APOSOR42282-250MG
Methyl 6-amino-4-methylnicotinate
Methyl 6-amino-4-methylnicotinate
Formel:
C₈H₁₀N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18803290 CAS-Nummer: 179555-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...