1-Chloro-9-iodo-phenazine

Lieferant: Apollo Scientific
Achtung

OR42290-1G OR42290-250MG OR42290-5G
APOSOR42290-1GEA 108 EUR
APOSOR42290-1G APOSOR42290-250MG APOSOR42290-5G
1-Chloro-9-iodo-phenazine
1-Chloro-9-iodo-phenazine
Formel: C₁₂H₆ClIN₂
Molecular Weight: 340.547 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32111680
CAS-Nummer: 2367002-82-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR