2-(2-Nitrovinyl)furan 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Fur-2-yl)-2-nitroethene, 1-(2-Furyl)-2-nitroethylene, 2-(2-Nitroethenyl)furan, 1-(Fur-2-yl)-2-nitroethylene

OR4271-100G OR4271-25G OR4271-5G
APOSOR4271-100GEA 886 EUR
APOSOR4271-100G APOSOR4271-25G APOSOR4271-5G
2-(2-Nitrovinyl)furan 98%
2-(2-Nitrovinyl)furan
Formel: C₆H₅NO₃
Molecular Weight: 139,109 g/mol
Schmelzpunkt: 72…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00030600
CAS-Nummer: 699-18-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR