N-Boc-1-Amino-1-cyclopentanemethanol
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl (1-(hydroxymethyl)cyclopentyl)carbamate
OR4311-1G
OR4311-250MG
APOSOR4311-1GEA
302
EUR
APOSOR4311-1G
APOSOR4311-250MG
N-Boc-1-Amino-1-cyclopentanemethanol
N-Boc-1-Amino-1-cyclopentanemethanol
Formel:
C₁₁H₂₁NO₃ Molecular Weight: 215.289 g/mol Schmelzpunkt: 108…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038539 CAS-Nummer: 168540-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...