7-Bromo-4-methyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR43500-1G OR43500-250MG
APOSOR43500-1GEA 864 EUR
APOSOR43500-1G APOSOR43500-250MG
7-Bromo-4-methyl-1H-indazole
7-Bromo-4-methyl-1H-indazole
Formel: C₈H₇BrN₂
Lagertemperatur: Kühlschrank
MDL: MFCD17168274
CAS-Nummer: 1257535-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR