7-Bromo-5-methyl-1H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
OR43501-1G
OR43501-250MG
OR43501-500MG
APOSOR43501-1GEA
85.7
EUR
APOSOR43501-1G
APOSOR43501-250MG
APOSOR43501-500MG
7-Bromo-5-methyl-1H-indazole
7-Bromo-5-methyl-1H-indazole
Formel:
C₈H₇BrN₂ Molecular Weight: 211.059 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD05664004 CAS-Nummer: 885272-97-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...