7-(Bromomethyl)isoquinoline hydrobromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 7-(Bromomethyl)-2-azanaphthalene hydrobromide

OR43505-100MG OR43505-1G OR43505-500MG
APOSOR43505-100MGEA 139 EUR
APOSOR43505-100MG APOSOR43505-1G APOSOR43505-500MG
7-(Bromomethyl)isoquinoline hydrobromide
7-(Bromomethyl)isoquinoline hydrobromide
Formel: C₁₀H₈BrN·BrH
Molecular Weight: 302.993 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12022338
CAS-Nummer: 1203372-02-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR