(2,6-Dichlorophenyl)methanesulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR43513-250MG
OR43513-1G
APOSOR43513-250MGEA
0
EUR
APOSOR43513-250MG
APOSOR43513-1G
(2,6-Dichlorophenyl)methanesulphonyl chloride
(2,6-Dichlorophenyl)methanesulphonyl chloride
Formel:
C₇H₅Cl₃O₂S Schmelzpunkt: 47…50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11204033 CAS-Nummer: 85952-31-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...