6-Aminopyridazin-3(2H)-one 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-1,6-dihydro-6-oxopyridazine, 6-Aminopyridazin-3-ol
OR43623-250MG
OR43623-1G
OR43623-5G
APOSOR43623-250MGEA
75.4
EUR
APOSOR43623-250MG
APOSOR43623-1G
APOSOR43623-5G
6-Aminopyridazin-3(2H)-one 97%
6-Aminopyridazin-3(2H)-one
Formel:
C₄H₅N₃O Molecular Weight: 111.102 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11520599 CAS-Nummer: 57041-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...