3-Amino-2-chloroisonicotinic acid 97%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-2-chloropyridine-4-carboxylic acid

OR43676-5G OR43676-1G
APOSOR43676-5GEA 690 EUR
APOSOR43676-5G APOSOR43676-1G
3-Amino-2-chloroisonicotinic acid 97%
3-Amino-2-chloroisonicotinic acid
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172.569 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15143308
CAS-Nummer: 58483-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR