3-Chloro-1H-indazole-6-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-1H-indazole-6-carbaldehyde

OR43678-1G OR43678-250MG OR43678-100MG
APOSOR43678-1GEA 606 EUR
APOSOR43678-1G APOSOR43678-250MG APOSOR43678-100MG
3-Chloro-1H-indazole-6-carboxaldehyde
3-Chloro-1H-indazole-6-carboxaldehyde
Formel: C₈H₅ClN₂O
Molecular Weight: 180,591 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11499021
CAS-Nummer: 1086391-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR