Methyl 3-chloro-4-(chlorosulphonyl)benzoate

Lieferant: Apollo Scientific
Gefahr

OR43682-1G OR43682-250MG OR43682-5G
APOSOR43682-1GEA 128 EUR
APOSOR43682-1G APOSOR43682-250MG APOSOR43682-5G
Methyl 3-chloro-4-(chlorosulphonyl)benzoate
Methyl 3-chloro-4-(chlorosulphonyl)benzoate
Formel: C₈H₆Cl₂O₄S
Molecular Weight: 269,102 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06637330
CAS-Nummer: 874830-93-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR