4-Bromo-2-chlorophenetole 98%
Lieferant: Apollo Scientific
Synonyme:
4-Bromo-2-chloro-ethoxybenzene, 4-Bromo-2-chloro-1-ethoxybenzene
OR43683-1G
OR43683-25G
OR43683-5G
APOSOR43683-1GEA
34.8
EUR
APOSOR43683-1G
APOSOR43683-25G
APOSOR43683-5G
4-Bromo-2-chlorophenetole 98%
4-Bromo-2-chlorophenetole
Formel:
C₈H₈BrClO Molecular Weight: 235.506 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09835099 CAS-Nummer: 279261-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...