1-(3-Chloro-4-iodophenyl)ethan-1-one 95%
Lieferant: Apollo Scientific
OR43694-1G
OR43694-250MG
APOSOR43694-1GEA
662
EUR
APOSOR43694-1G
APOSOR43694-250MG
1-(3-Chloro-4-iodophenyl)ethan-1-one 95%
1-(3-Chloro-4-iodophenyl)ethan-1-one
Formel:
C₈H₆ClIO Molecular Weight: 280,49 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28346645 CAS-Nummer: 149936-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...