2-Methyl-2H-indazole-3-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Carboxy-2-methyl-2H-indazole

OR4385-1G OR4385-250MG OR4385-5G
APOSOR4385-1GEA 94.8 EUR
APOSOR4385-1G APOSOR4385-250MG APOSOR4385-5G
2-Methyl-2H-indazole-3-carboxylic acid 98%
2-Methyl-2H-indazole-3-carboxylic acid
Formel: C₉H₈N₂O₂
Schmelzpunkt: 218…219 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368556
CAS-Nummer: 34252-44-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR