4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-[4-(Chlorocarbonyl)phenyl]-5-methyl-1,2,4-oxadiazole, 3-[4-(Chloroformyl)phenyl]-5-methyl-1,2,4-oxadiazole

OR4391-250MG OR4391-1G
APOSOR4391-250MGEA 0 EUR
APOSOR4391-250MG APOSOR4391-1G
4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoyl chloride 97%
4-(5-Methyl-1,2,4-oxadiazol-3-yl)benzoyl chloride
Formel: C₁₀H₇ClN₂O₂
Molecular Weight: 222.628 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368530
CAS-Nummer: 222541-76-6
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR