5-Bromo-1-methyl-1H-indole

Lieferant: Apollo Scientific
Gefahr

OR4414-1G OR4414-5G
APOSOR4414-1GEA 21.9 EUR
APOSOR4414-1G APOSOR4414-5G
5-Bromo-1-methyl-1H-indole
5-Bromo-1-methyl-1H-indole
Formel: C₉H₈BrN
Molecular Weight: 210,071 g/mol
Schmelzpunkt: 39…43 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00159857
CAS-Nummer: 10075-52-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR