4-Bromo-2,6-dimethylbenzenesulphonyl chloride 95%
Lieferant: Apollo Scientific
OR4497-1G
OR4497-250MG
APOSOR4497-1GEA
864
EUR
APOSOR4497-1G
APOSOR4497-250MG
4-Bromo-2,6-dimethylbenzenesulphonyl chloride 95%
4-Bromo-2,6-dimethylbenzenesulphonyl chloride
Formel:
C₈H₈BrClO₂S Molecular Weight: 283,57 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094653 CAS-Nummer: 351003-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...