6-(Benzyloxy)-1H-indazol-3-amine

Lieferant: Apollo Scientific

OR450010-1G OR450010-250MG
APOSOR450010-1GEA 242 EUR
APOSOR450010-1G APOSOR450010-250MG
6-(Benzyloxy)-1H-indazol-3-amine
6-(Benzyloxy)-1H-indazol-3-amine
Formel: C₁₄H₁₃N₃O
Molecular Weight: 239.273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845613
CAS-Nummer: 1167056-55-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR