3-Chloro-4-iodobenzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

OR450026-1G OR450026-250MG
APOSOR450026-1GEA 696 EUR
APOSOR450026-1G APOSOR450026-250MG
3-Chloro-4-iodobenzaldehyde 95%
3-Chloro-4-iodobenzaldehyde
Formel: C₇H₄ClIO
Molecular Weight: 266,464 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18393322
CAS-Nummer: 148334-14-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR