2-Thioxoimidazolidine-4,5-dione
Lieferant: Apollo Scientific
OR450093-1G
OR450093-250MG
APOSOR450093-1GEA
429
EUR
APOSOR450093-1G
APOSOR450093-250MG
2-Thioxoimidazolidine-4,5-dione
2-Thioxoimidazolidine-4,5-dione
Formel:
C₃H₂N₂O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00090436 CAS-Nummer: 496-89-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...