2-Amino-1,4-dihydroimidazol-5-one

Lieferant: Apollo Scientific

OR450094-1G OR450094-250MG
APOSOR450094-1GEA 429 EUR
APOSOR450094-1G APOSOR450094-250MG
2-Amino-1,4-dihydroimidazol-5-one
2-Amino-1,4-dihydroimidazol-5-one
Formel: C₃H₅N₃O
Molecular Weight: 99.091 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08444293
CAS-Nummer: 503-86-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR