5-bromo-4-chloro-thiazol-2-amine
Lieferant: Apollo Scientific
OR450104-1G
OR450104-250MG
APOSOR450104-1GEA
509
EUR
APOSOR450104-1G
APOSOR450104-250MG
5-bromo-4-chloro-thiazol-2-amine
5-bromo-4-chloro-thiazol-2-amine
Formel:
C₃H₂BrClN₂S Molecular Weight: 213.483 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24643186 CAS-Nummer: 1174132-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...