Ethyl 5-oxo-3-thioxo-2H-1,2,4-triazine-6-carboxylate

Lieferant: Apollo Scientific

OR450105-1G OR450105-250MG
APOSOR450105-1GEA 429 EUR
APOSOR450105-1G APOSOR450105-250MG
Ethyl 5-oxo-3-thioxo-2H-1,2,4-triazine-6-carboxylate
Ethyl 5-oxo-3-thioxo-2H-1,2,4-triazine-6-carboxylate
Formel: C₆H₇N₃O₃S
Molecular Weight: 201,203 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01567017
CAS-Nummer: 51101-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR