2-Iodothiazole

Lieferant: Apollo Scientific

OR450112-1G OR450112-250MG
APOSOR450112-1GEA 241 EUR
APOSOR450112-1G APOSOR450112-250MG
2-Iodothiazole
2-Iodothiazole
Formel: C₃H₂INS
Molecular Weight: 211.024 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02094167
CAS-Nummer: 3034-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR