α-Tripiperidine
Lieferant: Apollo Scientific
OR450113-1G
OR450113-250MG
APOSOR450113-1GEA
683
EUR
APOSOR450113-1G
APOSOR450113-250MG
α-Tripiperidine
α-Tripiperidine
Formel:
C₁₅H₂₇N₃ Molecular Weight: 249.395 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18839150 CAS-Nummer: 522-33-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...