3-Nitrobenzoyl isocyanate
Lieferant: Apollo Scientific
OR450134-1G
OR450134-250MG
APOSOR450134-1GEA
509
EUR
APOSOR450134-1G
APOSOR450134-250MG
3-Nitrobenzoyl isocyanate
3-Nitrobenzoyl isocyanate
May form small amount of sediment
Formel:
C₈H₄N₂O₄ Molecular Weight: 192,128 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD28365515 CAS-Nummer: 5843-49-2 UN: 2206 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...