2-Nitrobenzoyl isocyanate

Lieferant: Apollo Scientific

OR450135-1G OR450135-250MG
APOSOR450135-1GEA 429 EUR
APOSOR450135-1G APOSOR450135-250MG
2-Nitrobenzoyl isocyanate
2-Nitrobenzoyl isocyanate

May form small amount of sediment

Formel: C₈H₄N₂O₄
Molecular Weight: 192,128 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD32264081
CAS-Nummer: 5843-48-1
UN: 2206
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR