3-Iodo-1H-indole-5-carbonitrile

Lieferant: Apollo Scientific

OR45089-10G OR45089-1G OR45089-5G
APOSOR45089-10GEA 525 EUR
APOSOR45089-10G APOSOR45089-1G APOSOR45089-5G
3-Iodo-1H-indole-5-carbonitrile
3-Iodo-1H-indole-5-carbonitrile
Formel: C₉H₅IN₂
Molecular Weight: 268,054 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22683211
CAS-Nummer: 1092114-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR