3-Azetidineacetic acid hydrochloride

Lieferant: Apollo Scientific

OR451017-1G OR451017-250MG
APOSOR451017-1GEA 406 EUR
APOSOR451017-1G APOSOR451017-250MG
3-Azetidineacetic acid hydrochloride
3-Azetidineacetic acid hydrochloride
Formel: C₅H₉NO₂·ClH
Molecular Weight: 151,591 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11867905
CAS-Nummer: 952675-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR