1-(3-Hydroxy-1-azetidinyl)-ethanone 95%
Lieferant: Apollo Scientific
Synonyme:
1-(3-Hydroxyazetidin-1-yl)ethanone
OR451031-1G
OR451031-10G
OR451031-5G
APOSOR451031-1GEA
168
EUR
APOSOR451031-1G
APOSOR451031-10G
APOSOR451031-5G
1-(3-Hydroxy-1-azetidinyl)-ethanone 95%
1-(3-Hydroxy-1-azetidinyl)-ethanone
Formel:
C₅H₉NO₂ Molecular Weight: 115,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07366667 CAS-Nummer: 118972-96-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...