1-(3-Bromophenyl)-cyclobutanemethanamine 95%

Lieferant: Apollo Scientific

Synonyme: (1-(3-Bromophenyl)cyclobutyl)methanamine

OR451073-1G OR451073-250MG
APOSOR451073-1GEA 543 EUR
APOSOR451073-1G APOSOR451073-250MG
1-(3-Bromophenyl)-cyclobutanemethanamine 95%
1-(3-Bromophenyl)-cyclobutanemethanamine
Formel: C₁₁H₁₄BrN
Molecular Weight: 240,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11642693
CAS-Nummer: 915690-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR