(R)-4-Benzyl-2-hydroxymethylpiperazine

Lieferant: Apollo Scientific

OR451103-1G OR451103-250MG
APOSOR451103-1GEA 190 EUR
APOSOR451103-1G APOSOR451103-250MG
(R)-4-Benzyl-2-hydroxymethylpiperazine
(R)-4-Benzyl-2-hydroxymethylpiperazine
Formel: C₁₂H₁₈N₂O
Molecular Weight: 206,284 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11113046
CAS-Nummer: 149715-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR