1-[(Boc-amino)methyl]-cyclobutanecarboxylic acid 98%

Lieferant: Apollo Scientific

Synonyme: 1-(((tert-Butoxycarbonyl)amino)methyl)cyclobutane-1-carboxylic acid

OR451122-1G OR451122-5G
APOSOR451122-1GEA 241 EUR
APOSOR451122-1G APOSOR451122-5G
1-[(Boc-amino)methyl]-cyclobutanecarboxylic acid 98%
1-[(Boc-amino)methyl]-cyclobutanecarboxylic acid
Formel: C₁₁H₁₉NO₄
Molecular Weight: 229,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20485946
CAS-Nummer: 220145-21-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR