3-Iodo-2-nitroanisole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Iodo-3-methoxy-2-nitrobenzene, 3-Iodo-2-nitrophenyl methyl ether

OR45118-1G OR45118-250MG OR45118-5G
APOSOR45118-1GEA 186 EUR
APOSOR45118-1G APOSOR45118-250MG APOSOR45118-5G
3-Iodo-2-nitroanisole
3-Iodo-2-nitroanisole
Formel: C₇H₆INO₃
Molecular Weight: 279,032 g/mol
Schmelzpunkt: 81…83 °C
Lagertemperatur: Kühlschrank
MDL: MFCD18397933
CAS-Nummer: 725266-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR