4-Bromo-2-iodoanisole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-iodo-1-methoxybenzene, 4-Bromo-2-iodophenyl methyl ether

OR45119-1G OR45119-250MG OR45119-5G
APOSOR45119-1GEA 29 EUR
APOSOR45119-1G APOSOR45119-250MG APOSOR45119-5G
4-Bromo-2-iodoanisole
4-Bromo-2-iodoanisole
Formel: C₇H₆BrIO
Molecular Weight: 312.93 g/mol
Schmelzpunkt: 64…68 °C
Lagertemperatur: Kühlschrank
MDL: MFCD13186707
CAS-Nummer: 98273-59-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR