2-Bromo-6-iodoanisole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-3-iodo-2-methoxybenzene, 2-Bromo-6-iodophenyl methyl ether

OR45123-1G OR45123-250MG
APOSOR45123-1GEA 140 EUR
APOSOR45123-1G APOSOR45123-250MG
2-Bromo-6-iodoanisole
2-Bromo-6-iodoanisole
Formel: C₇H₆BrIO
Molecular Weight: 312,93 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18391617
CAS-Nummer: 860584-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR