4-(2-Bromoethyl)phenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Hydroxyphenethyl bromide
OR45134-25G
OR45134-5G
OR45134-10G
APOSOR45134-25GEA
212
EUR
APOSOR45134-25G
APOSOR45134-5G
APOSOR45134-10G
4-(2-Bromoethyl)phenol 97%
4-(2-Bromoethyl)phenol
Formel:
C₈H₉BrO Molecular Weight: 201,06 g/mol Schmelzpunkt: 88…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129679 CAS-Nummer: 14140-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...