4-Bromo-1-(chloromethyl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR451403-1G OR451403-5G
APOSOR451403-1GEA 377 EUR
APOSOR451403-1G APOSOR451403-5G
4-Bromo-1-(chloromethyl)-1H-pyrazole
4-Bromo-1-(chloromethyl)-1H-pyrazole
Formel: C₄H₄BrClN₂
Molecular Weight: 195,445 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03423393
CAS-Nummer: 861001-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR