4-Chloro-5-methyl-1H-pyrazol-3-amine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR451405-1G
OR451405-5G
APOSOR451405-1GEA
31.9
EUR
APOSOR451405-1G
APOSOR451405-5G
4-Chloro-5-methyl-1H-pyrazol-3-amine hydrochloride
4-Chloro-5-methyl-1H-pyrazol-3-amine hydrochloride
Formel:
C₄H₆ClN₃·ClH Molecular Weight: 168,024 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00767237 CAS-Nummer: 130128-49-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...