2-(4-Bromo-1H-pyrazol-1-yl)ethanamine hydrochloride

Lieferant: Apollo Scientific
Gefahr

OR451407-1G OR451407-5G
APOSOR451407-1GEA 36.4 EUR
APOSOR451407-1G APOSOR451407-5G
2-(4-Bromo-1H-pyrazol-1-yl)ethanamine hydrochloride
2-(4-Bromo-1H-pyrazol-1-yl)ethanamine hydrochloride
Formel: C₅H₈BrN₃·ClH
Molecular Weight: 226.502 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04967288
CAS-Nummer: 663941-75-1
UN: 3259
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR