4-Chloro-5-cyclopropyl-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR451408-1G OR451408-5G
APOSOR451408-1GEA 33.5 EUR
APOSOR451408-1G APOSOR451408-5G
4-Chloro-5-cyclopropyl-1H-pyrazole
4-Chloro-5-cyclopropyl-1H-pyrazole
Formel: C₆H₇ClN₂
Molecular Weight: 142.586 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12548010
CAS-Nummer: 1248516-15-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR