(3-Cyclopropylisoxazol-5-yl)methanol 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR451451-1G
OR451451-250MG
APOSOR451451-1GEA
197
EUR
APOSOR451451-1G
APOSOR451451-250MG
(3-Cyclopropylisoxazol-5-yl)methanol 95%
(3-Cyclopropylisoxazol-5-yl)methanol
Formel:
C₇H₉NO₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD16556130 CAS-Nummer: 121604-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...